missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NA-Star™ Influenza Neuraminidase Inhibitor Resistance Detection Kit

Catalog No. 4374422
Encompass
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4374422 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 4374422 Supplier Invitrogen™ Supplier No. 4374422

Includes

Substrate, required buffer and reaction accelerator

Rapid and sensitive quantitation of influenza neuraminidase inhibitor resistance in dilutions of virus culture medium in a 96-well microplate format. The NA-Star Influenza Neuraminidase Inhibitor Resistance Detection Kit includes everything needed to qua

NA-Star Influenza Neuraminidase Inhibitor Resistance Detection Reagent Set includes everything needed to quantitate neuraminidase activity and neuraminidase inhibitor resistance in avian, equine, human (types A and B), and porcine influenza viruses. The kit's fast and easy protocol and convenient 96-well plate format make it ideal for monitoring influenza virus neuraminidase inhibitor resistance, as well as high-throughput inhibitor compound screening.

  • Includes pre-made solutions for all required reagents, enabling you to standardize assays and compare results between labs easily
  • Provides up to 50-fold higher sensitivity than assays using fluorescent methylumbelliferone N-acetylneuraminic acid (MUNANA) substrate
  • Dynamic range of four orders of magnitude enables you to quantitate virus isolates over a broad range of virus concentration and with varying levels of neuraminidase activity without performing numerous sample dilutions
  • Fast and easy protocol enables you to complete assays in less than 1.5 hours, providing rapid assay throughput

Specifications

  • Target: Neuraminidase
  • Detection Method: Enhanced Chemiluminescence
  • For use with Equipment: Luminometer (Microplate)
  • Form: Liquid
  • High Throughput Compatibility: High Throughput-Compatible
  • Readout: End Point
  • Substrate Properties: Chemical Substrate
  • Substrate Type: Neuraminidase Substrate

Specifications

Content And Storage This kit contains the enzyme substrate, assay buffer, reaction accelerator, and microplates. Store reagent set components at 2°C to 8°C.
Detection Method Chemiluminescence
For Use With (Application) Microbiological Analysis
For Use With (Equipment) Luminometer (Microplate)
Product Type Neuraminidase Assay
Form Liquid
Format 96-well plate
Product Line NA-Star, NovaBright
Quantity 1 kit
Substrate NA-Star™
Substrate Properties Chemical Substrate
Substrate Type Neuraminidase Substrate
Target Enzyme Sialidase, Neuraminidase
Throughput High-throughput Compatible
Validated Application Enzyme Assay
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.