missing translation for 'onlineSavingsMsg'
Learn More

N6-methyladenosine (m6A) Rabbit anti-All, Polyclonal, MilliporeSigma™

Catalog No. ABE572I25UG
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABE572I25UG 25 μg
ABE572I100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ABE572I25UG Supplier MilliporeSigma Supplier No. ABE572I25UG
Only null left

Rabbit Polyclonal Antibody

N6-methyladenosine (m6A) Polyclonal antibody specifically detects N6-methyladenosine (m6A) in All samples. It is validated for ChIP assay (ChIP), Immunoprecipitation, Western Blotting.

Specifications

Antigen N6-methyladenosine (m6A)
Applications ChIP Assay, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in PBS with 0.05% sodium azide.
Host Species Rabbit
Immunogen BSA-conjugated N6-methyladenosine
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species All species
Content And Storage Stable for 1 year at +2°C to +8°C from date of receipt.
Form Purified
Isotype IgG
Show More Show Less