missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N6-Benzyladenosine

Catalog No. ac226420010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC226420010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC226420010 Supplier Thermo Scientific Chemicals Supplier No. 226420010
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

CAS 4294-16-0
CAS Min % 99.0
CAS Max % 100.0
Color White
Elemental Analysis 99.0% min.
Infrared Spectrum Authentic
Assay Percent Range 99+%
Molecular Formula C17H19N5O4
Quantity 1 g
MDL Number MFCD00005740
Beilstein 26, IV, 3682
Synonym n6-benzyladenosine, 6-benzylaminopurine riboside, unii-0m6s0fah3t, n-benzyladenosine, 0m6s0fah3t, benzyladenosine, 6-benzyladenosine, 2r,3r,4s,5r-2-6-benzylamino-9h-purin-9-yl-5-hydroxymethyl tetrahydrofuran-3,4-diol, 2r,3r,4s,5r-2-6-benzylamino purin-9-yl-5-hydroxymethyl oxolane-3,4-diol, n6-benzyl adenosine
InChI Key MRPKNNSABYPGBF-ABJKBEJCNA-N
SMILES OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N1C=NC2=C(NCC3=CC=CC=C3)N=CN=C12
IUPAC Name (2R,3R,4S,5R)-2-[6-(benzylamino)-9H-purin-9-yl]-5-(hydroxymethyl)oxolane-3,4-diol
Molecular Weight (g/mol) 357.37
PubChem CID 92208
Formula Weight 357.36
Physical Form Powder
Chemical Name or Material 6-Benzylaminopurine riboside, 99+%
Show More Show Less
EINECSNumber 224-298-9

RUO – Research Use Only