missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N6-Benzoyladenine 98.0+%, TCI America™
Supplier: TCI America B33445G
Specifications
| N6-Benzoyladenine | |
| 242°C | |
| C12H9N5O | |
| 5 g | |
| QQJXZVKXNSFHRI-UHFFFAOYSA-N | |
| N-(7H-purin-6-yl)benzamide | |
| 97075 | |
| ≥98.0% (HPLC,T) |
| 4005-49-6 | |
| White | |
| MFCD00037927 | |
| n6-benzoyladenine, n-7h-purin-6-yl benzamide, n-benzoylaminopurine, n-9h-purin-6-yl benzamide, 6-benzoylaminopurine, benzamide, n-1h-purin-6-yl, 6-benzamidopurine, adenine,6n-benzoyl, phenyl-n-purin-6-ylcarboxamide, n-benzoyladenine | |
| C1=CC=C(C=C1)C(=O)NC2=NC=NC3=C2NC=N3 | |
| 239.238 | |
| 239.24 | |
| Crystalline Powder |
Chemical Identifiers
| 4005-49-6 | |
| 239.238 | |
| QQJXZVKXNSFHRI-UHFFFAOYSA-N | |
| 97075 | |
| C1=CC=C(C=C1)C(=O)NC2=NC=NC3=C2NC=N3 |
| C12H9N5O | |
| MFCD00037927 | |
| n6-benzoyladenine, n-7h-purin-6-yl benzamide, n-benzoylaminopurine, n-9h-purin-6-yl benzamide, 6-benzoylaminopurine, benzamide, n-1h-purin-6-yl, 6-benzamidopurine, adenine,6n-benzoyl, phenyl-n-purin-6-ylcarboxamide, n-benzoyladenine | |
| N-(7H-purin-6-yl)benzamide |
Safety and Handling
TSCA : No