accessibility menu, dialog, popup

UserName

CAS RN 4005-49-6

IUPAC Name: N-(7H-purin-6-yl)benzamide
Synonyms: N-(7H-purin-6-yl)benzamide
Molecular Weight (g/mol): 239.24
Molecular Formula: C12H9N5O
InChi Key: QQJXZVKXNSFHRI-UHFFFAOYSA-N
SMILES: O=C(NC1=C2NC=NC2=NC=N1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)