missing translation for 'onlineSavingsMsg'
Learn More

N6-Benzoyladenine 98.0+%, TCI America™

Catalog No. b334425g
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B334425G 25 g
B33445G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B334425G Supplier TCI America Supplier No. B334425G
Only null left

Chemical Identifiers

CAS 4005-49-6
Molecular Formula C12H9N5O
Molecular Weight (g/mol) 239.238
MDL Number MFCD00037927
InChI Key QQJXZVKXNSFHRI-UHFFFAOYSA-N
Synonym n6-benzoyladenine, n-7h-purin-6-yl benzamide, n-benzoylaminopurine, n-9h-purin-6-yl benzamide, 6-benzoylaminopurine, benzamide, n-1h-purin-6-yl, 6-benzamidopurine, adenine,6n-benzoyl, phenyl-n-purin-6-ylcarboxamide, n-benzoyladenine
PubChem CID 97075
IUPAC Name N-(7H-purin-6-yl)benzamide
SMILES C1=CC=C(C=C1)C(=O)NC2=NC=NC3=C2NC=N3

Specifications

Melting Point 242°C
Color White
Quantity 25 g
Formula Weight 239.24
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N6-Benzoyladenine