missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N6-Benzoyl-5'-O-(4,4'-dimethoxytrityl)-2'-deoxyadenosine 99.0+%, TCI America™
$138.61 - $556.38
Chemical Identifiers
| CAS | 64325-78-6 |
|---|---|
| Molecular Formula | C38H35N5O6 |
| Molecular Weight (g/mol) | 657.73 |
| MDL Number | MFCD00010058 |
| InChI Key | LPICNYATEWGYHI-WIHCDAFUSA-N |
| PubChem CID | 2724489 |
| IUPAC Name | N-{9-[(2R,4S,5R)-5-{[bis(4-methoxyphenyl)(phenyl)methoxy]methyl}-4-hydroxyoxolan-2-yl]-9H-purin-6-yl}benzamide |
| SMILES | COC1=CC=C(C=C1)C(OC[C@H]1O[C@H](C[C@@H]1O)N1C=NC2=C(NC(=O)C3=CC=CC=C3)N=CN=C12)(C1=CC=CC=C1)C1=CC=C(OC)C=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
B3103100MG
|
TCI America
B3103100MG |
100 mg |
Each for $138.61
|
|
|||||
|
B31031G
|
TCI America
B31031G |
1 g |
Each for $556.38
|
|
|||||
Chemical Identifiers
| 64325-78-6 | |
| 657.73 | |
| LPICNYATEWGYHI-WIHCDAFUSA-N | |
| N-{9-[(2R,4S,5R)-5-{[bis(4-methoxyphenyl)(phenyl)methoxy]methyl}-4-hydroxyoxolan-2-yl]-9H-purin-6-yl}benzamide |
| C38H35N5O6 | |
| MFCD00010058 | |
| 2724489 | |
| COC1=CC=C(C=C1)C(OC[C@H]1O[C@H](C[C@@H]1O)N1C=NC2=C(NC(=O)C3=CC=CC=C3)N=CN=C12)(C1=CC=CC=C1)C1=CC=C(OC)C=C1 |
Specifications
| 64325-78-6 | |
| White | |
| MFCD00010058 | |
| n6-benzoyl-5'-o-4,4'-dimethoxytrityl-2'-deoxyadenosine, dmbta, bz-dmt-da, n 6-benzoyl-5'-o-4,4'-dimethoxytrityl-2'-deoxyadenosine, adenosine, n-benzoyl-5'-o-bis 4-methoxyphenyl phenylmethyl-2'-deoxy, n-9-2r,4s,5r-5-bis 4-methoxyphenyl phenyl methoxy methyl-4-hydroxyoxolan-2-yl purin-6-yl benzamide, ksc621a9d, n-benzoyl-5'-o-bis 4-methoxyphenyl phenylmethyl-2'-deoxyadenosine, 5'-ortho-dmt-n6-bz-2'-deoxyadenosine | |
| COC1=CC=C(C=C1)C(OC[C@H]1O[C@H](C[C@@H]1O)N1C=NC2=C(NC(=O)C3=CC=CC=C3)N=CN=C12)(C1=CC=CC=C1)C1=CC=C(OC)C=C1 | |
| 657.73 | |
| 657.73 | |
| Crystalline Powder |
| 94°C | |
| C38H35N5O6 | |
| 100 mg | |
| LPICNYATEWGYHI-WIHCDAFUSA-N | |
| N-{9-[(2R,4S,5R)-5-{[bis(4-methoxyphenyl)(phenyl)methoxy]methyl}-4-hydroxyoxolan-2-yl]-9H-purin-6-yl}benzamide | |
| 2724489 | |
| ≥99.0% (HPLC) | |
| N6-Benzoyl-5′-O-(4,4′-dimethoxytrityl)-2′-deoxyadenosine |
Safety and Handling
TSCA : Yes