missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N4-Benzoyl-3',5'-O-(1,1,3,3-tetraisopropyl-1,3-disiloxanediyl)cytidine 97.0+%, TCI America™
$379.72 - $1313.84
Chemical Identifiers
| CAS | 69304-43-4 |
|---|---|
| Molecular Formula | C28H43N3O7Si2 |
| Molecular Weight (g/mol) | 589.836 |
| InChI Key | NFIAUHQTSCWVMS-LYPBTDJXSA-N |
| PubChem CID | 10054127 |
| IUPAC Name | N-[1-[(6aR,8R,9R,9aS)-9-hydroxy-2,2,4,4-tetra(propan-2-yl)-6a,8,9,9a-tetrahydro-6H-furo[3,2-f][1,3,5,2,4]trioxadisilocin-8-yl]-2-oxopyrimidin-4-yl]benzamide |
| SMILES | CC(C)[Si]1(OCC2C(C(C(O2)N3C=CC(=NC3=O)NC(=O)C4=CC=CC=C4)O)O[Si](O1)(C(C)C)C(C)C)C(C)C |
Chemical Identifiers
| 69304-43-4 | |
| 589.836 | |
| 10054127 | |
| CC(C)[Si]1(OCC2C(C(C(O2)N3C=CC(=NC3=O)NC(=O)C4=CC=CC=C4)O)O[Si](O1)(C(C)C)C(C)C)C(C)C |
| C28H43N3O7Si2 | |
| NFIAUHQTSCWVMS-LYPBTDJXSA-N | |
| N-[1-[(6aR,8R,9R,9aS)-9-hydroxy-2,2,4,4-tetra(propan-2-yl)-6a,8,9,9a-tetrahydro-6H-furo[3,2-f][1,3,5,2,4]trioxadisilocin-8-yl]-2-oxopyrimidin-4-yl]benzamide |
Specifications
| 69304-43-4 | |
| White-Yellow | |
| 1 g | |
| CC(C)[Si]1(OCC2C(C(C(O2)N3C=CC(=NC3=O)NC(=O)C4=CC=CC=C4)O)O[Si](O1)(C(C)C)C(C)C)C(C)C | |
| 589.836 | |
| 589.84 | |
| Crystalline Powder |
| 101°C | |
| C28H43N3O7Si2 | |
| NFIAUHQTSCWVMS-LYPBTDJXSA-N | |
| N-[1-[(6aR,8R,9R,9aS)-9-hydroxy-2,2,4,4-tetra(propan-2-yl)-6a,8,9,9a-tetrahydro-6H-furo[3,2-f][1,3,5,2,4]trioxadisilocin-8-yl]-2-oxopyrimidin-4-yl]benzamide | |
| 10054127 | |
| ≥97.0% (HPLC,N) | |
| N4-Benzoyl-3′,5′-O-(1,1,3,3-tetraisopropyl-1,3-disiloxanediyl)cytidine |
Safety and Handling
TSCA : No