missing translation for 'onlineSavingsMsg'
Learn More

N4-Benzoyl-2'-deoxycytidine 98.0+%, TCI America™

Catalog No. B31021G
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B31021G 1 g
B3102100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B31021G Supplier TCI America Supplier No. B31021G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4836-13-9
Molecular Formula C16H17N3O5
Molecular Weight (g/mol) 331.33
MDL Number MFCD00010115
InChI Key MPSJHJFNKMUKCN-OUCADQQQSA-N
Synonym n4-benzoyl-2'-deoxycytidine, n-benzoyl-2'-deoxy-cytidine, n-benzoyl-2'-deoxycytidine, cytidine, n-benzoyl-2'-deoxy, cytidine,n-benzoyl-2'-deoxy, n-benzoyl-2-deoxy-cytidine, dc-bz, ksc590o7t, n-benzoyl-2/'-deoxycytidine
PubChem CID 9797617
IUPAC Name N-{1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-2-oxo-1,2-dihydropyrimidin-4-yl}benzamide
SMILES OC[C@H]1O[C@H](C[C@@H]1O)N1C=CC(NC(=O)C2=CC=CC=C2)=NC1=O

Specifications

Melting Point 207°C
Color White-Yellow
Quantity 1 g
Formula Weight 331.33
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material N4-Benzoyl-2′-deoxycytidine