accessibility menu, dialog, popup

UserName

CAS RN 4836-13-9

IUPAC Name: N-{1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-2-oxo-1,2-dihydropyrimidin-4-yl}benzamide
Synonyms: cytidine, N-benzoyl-2'-deoxy-
Molecular Weight (g/mol): 331.33
Molecular Formula: C16H17N3O5
InChi Key: MPSJHJFNKMUKCN-OUCADQQQSA-N
SMILES: OC[C@H]1O[C@H](C[C@@H]1O)N1C=CC(NC(=O)C2=CC=CC=C2)=NC1=O
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)