missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N4-Acetylsulfamethoxazole, ≥98.5% (HPLC), MilliporeSigma™ Supelco™
Description
Pharmaceutical (small molecule)
Specifications
Specifications
| CAS | 21312-10-7 |
| Physical Form | Neat |
| Linear Formula | C12H13N3O4 S |
| MDL Number | MFCD00205417 |
| Synonym | N-{4-{[(5-Methyl-3-isoxazolyl)amino]sulfonyl}phenyl}acetamide; Sulfisomezole-N4-acetate |
| Type | Analytical Standard |
| Quantity | 25 mg |
| Formula Weight | 295.31 |
| Percent Purity | ≥98.5% (HPLC) |
| Grade | Analytical Standard |