missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N1-Methyl-4-nitro-1,2-phenylenediamine 98.0+%, TCI America™
$42.36 - $102.03
Chemical Identifiers
| CAS | 41939-61-1 |
|---|---|
| Molecular Formula | C7H9N3O2 |
| Molecular Weight (g/mol) | 167.17 |
| MDL Number | MFCD00156607 |
| InChI Key | MNIKERWISBANET-UHFFFAOYSA-N |
| Synonym | 1,2-Diamino-N1-methyl-4-nitrobenzene, 2-Amino-1-methylamino-4-nitrobenzene |
| PubChem CID | 3420025 |
| IUPAC Name | N1-methyl-4-nitrobenzene-1,2-diamine |
| SMILES | CNC1=C(N)C=C(C=C1)[N+]([O-])=O |
Chemical Identifiers
| 41939-61-1 | |
| 167.17 | |
| MNIKERWISBANET-UHFFFAOYSA-N | |
| 3420025 | |
| CNC1=C(N)C=C(C=C1)[N+]([O-])=O |
| C7H9N3O2 | |
| MFCD00156607 | |
| 1,2-Diamino-N1-methyl-4-nitrobenzene, 2-Amino-1-methylamino-4-nitrobenzene | |
| N1-methyl-4-nitrobenzene-1,2-diamine |
Specifications
| 41939-61-1 | |
| Red | |
| MFCD00156607 | |
| 2811 | |
| MNIKERWISBANET-UHFFFAOYSA-N | |
| N1-methyl-4-nitrobenzene-1,2-diamine | |
| 3420025 | |
| ≥98.0% (GC) | |
| N1-Methyl-4-nitro-1,2-phenylenediamine |
| 179°C | |
| C7H9N3O2 | |
| 1 g | |
| 1,2-Diamino-N1-methyl-4-nitrobenzene, 2-Amino-1-methylamino-4-nitrobenzene | |
| CNC1=C(N)C=C(C=C1)[N+]([O-])=O | |
| 167.17 | |
| 167.17 | |
| Crystalline Powder |
Safety and Handling
TSCA : No