missing translation for 'onlineSavingsMsg'
Learn More

N1-Methyl-4-nitro-1,2-phenylenediamine 98.0+%, TCI America™

Catalog No. M25231G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M25231G 1 g
M25235G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M25231G Supplier TCI America Supplier No. M25231G
Only null left

Chemical Identifiers

CAS 41939-61-1
Molecular Formula C7H9N3O2
Molecular Weight (g/mol) 167.17
MDL Number MFCD00156607
InChI Key MNIKERWISBANET-UHFFFAOYSA-N
Synonym 1,2-Diamino-N1-methyl-4-nitrobenzene, 2-Amino-1-methylamino-4-nitrobenzene
PubChem CID 3420025
IUPAC Name N1-methyl-4-nitrobenzene-1,2-diamine
SMILES CNC1=C(N)C=C(C=C1)[N+]([O-])=O

Specifications

Melting Point 179°C
Color Red
Quantity 1 g
UN Number 2811
Formula Weight 167.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N1-Methyl-4-nitro-1,2-phenylenediamine