missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N^w-Nitro-L-arginine, 98%
CAS: 2149-70-4 | C6H13N5O4 | 219.201 g/mol
$106.19 - $327.30
Chemical Identifiers
| CAS | 2149-70-4 |
|---|---|
| Molecular Formula | C6H13N5O4 |
| Molecular Weight (g/mol) | 219.201 |
| MDL Number | MFCD00007033 |
| InChI Key | MRAUNPAHJZDYCK-BYPYZUCNSA-N |
| Synonym | h-arg no2-oh, ng-nitro-l-arginine, l-nna, nitroarginine, n-nitro-l-arginine, nomega-nitro-l-arginine, n g-nitro-l-arginine, ng-no2-l-arg, n-omega-nitro-l-arginine, n g-nitroarginine |
| PubChem CID | 440005 |
| ChEBI | CHEBI:27960 |
| SMILES | C(CC(C(=O)O)N)CN=C(N)N[N+](=O)[O-] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6206106
|
Thermo Scientific Chemicals
H6206106 |
5 g |
Each for $106.19
|
|
|||||
|
AAH6206114
|
Thermo Scientific Chemicals
H6206114 |
25 g |
Each for $327.30
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 2149-70-4 | |
| 219.201 | |
| MRAUNPAHJZDYCK-BYPYZUCNSA-N | |
| 440005 | |
| C(CC(C(=O)O)N)CN=C(N)N[N+](=O)[O-] |
| C6H13N5O4 | |
| MFCD00007033 | |
| h-arg no2-oh, ng-nitro-l-arginine, l-nna, nitroarginine, n-nitro-l-arginine, nomega-nitro-l-arginine, n g-nitro-l-arginine, ng-no2-l-arg, n-omega-nitro-l-arginine, n g-nitroarginine | |
| CHEBI:27960 |
Specifications
| 257°C | |
| C6H13N5O4 | |
| 1728914 | |
| MRAUNPAHJZDYCK-BYPYZUCNSA-N | |
| 5 g | |
| 440005 | |
| 219.2 | |
| Nomega-Nitro-L-arginine |
| 2149-70-4 | |
| MFCD00007033 | |
| h-arg no2-oh, ng-nitro-l-arginine, l-nna, nitroarginine, n-nitro-l-arginine, nomega-nitro-l-arginine, n g-nitro-l-arginine, ng-no2-l-arg, n-omega-nitro-l-arginine, n g-nitroarginine | |
| C(CC(C(=O)O)N)CN=C(N)N[N+](=O)[O-] | |
| 219.201 | |
| CHEBI:27960 | |
| 98% |
Safety and Handling
EINECSNumber : 218-418-9
RTECSNumber : RM2982100
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only