missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Vinylacetamide 98.0+%, TCI America™
$167.49 - $498.62
Chemical Identifiers
| CAS | 5202-78-8 |
|---|---|
| Molecular Formula | C4H7NO |
| Molecular Weight (g/mol) | 85.106 |
| MDL Number | MFCD00081205 |
| InChI Key | RQAKESSLMFZVMC-UHFFFAOYSA-N |
| PubChem CID | 78875 |
| IUPAC Name | N-ethenylacetamide |
| SMILES | CC(=O)NC=C |
Chemical Identifiers
| 5202-78-8 | |
| 85.106 | |
| RQAKESSLMFZVMC-UHFFFAOYSA-N | |
| N-ethenylacetamide |
| C4H7NO | |
| MFCD00081205 | |
| 78875 | |
| CC(=O)NC=C |
Specifications
| 5202-78-8 | |
| White-Yellow | |
| C4H7NO | |
| 5 g | |
| CC(=O)NC=C | |
| 85.106 | |
| 85.11 | |
| Crystalline Powder |
| 55°C | |
| 96°C | |
| MFCD00081205 | |
| RQAKESSLMFZVMC-UHFFFAOYSA-N | |
| N-ethenylacetamide | |
| 78875 | |
| ≥98.0% (GC) | |
| N-Vinylacetamide |
Safety and Handling
EINECSNumber : (2)-3701
TSCA : Yes