accessibility menu, dialog, popup

UserName

CAS RN 5202-78-8

CH3OHNH2C

CAS RN 5202-78-8

IUPAC Name: N-ethenylacetamide
Synonyms: N-vinylacetamide poly(N-vinylacetamide)
Molecular Weight (g/mol): 85.11
Molecular Formula: C4H7NO
InChi Key: RQAKESSLMFZVMC-UHFFFAOYSA-N
SMILES: CC(=O)NC=C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results