missing translation for 'onlineSavingsMsg'
Learn More

N-Vinyl-epsilon-caprolactam (stabilized with HO-TEMPO) 98.0+%, TCI America™

Catalog No. p-7137914
Change view
Click to view available options
Quantity:
100 g
500 g
Catalog No. Quantity
V0073100G 100 g
V0073500G 500 g
2 options

Catalog No. V0073100G

Supplier: TCI America V0073100G

Only null left

Chemical Identifiers

CAS 2235-00-9
Molecular Formula C8H13NO
Molecular Weight (g/mol) 139.198
MDL Number MFCD00080693
InChI Key JWYVGKFDLWWQJX-UHFFFAOYSA-N
Synonym n-vinylcaprolactam, 1-vinylazepan-2-one, n-vinyl-epsilon-caprolactam, 2h-azepin-2-one, 1-ethenylhexahydro, unii-kfc10cy9up, 1-vinylhexahydro-2h-azepin-2-one, 1-ethenylhexahydro-2h-azepin-2-one, kfc10cy9up, 1-vinylazaperhydroepin-2-one, poly-n-vinylcaprolactam
PubChem CID 75227
IUPAC Name 1-ethenylazepan-2-one
SMILES C=CN1CCCCCC1=O

Specifications

Melting Point 36°C
Color White-Yellow
Quantity 100 g
Formula Weight 139.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material N-Vinyl-epsilon-caprolactam (stabilized with HO-TEMPO)
EINECSNumber (5)-6239
TSCA Yes