missing translation for 'onlineSavingsMsg'
Learn More

N-Vinyl-epsilon-caprolactam, 99%

Catalog No. AAB2131109
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2131109 10 g
AAB2131118 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2131109 Supplier Thermo Scientific Chemicals Supplier No. B2131109
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2235-00-9 | C8H13NO | 139.198 g/mol

N-Vinyl-epsilon-caprolactam is used as reacive diluent in a number of our screen and is used in a wide range of UV curing screen printing inks. N-Vinyl-epsilon-caprolactam is used in UV-coatings and -adhesives as well as a building block for synthesis of paper coatings. Reactive diluent for in optical fiber coatings and rapid prototyping.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Vinyl-epsilon-caprolactam is used as reacive diluent in a number of our screen and is used in a wide range of UV curing screen printing inks. N-Vinyl-epsilon-caprolactam is used in UV-coatings and -adhesives as well as a building block for synthesis of paper coatings. Reactive diluent for in optical fiber coatings and rapid prototyping.

Solubility
Soluble in water (partly).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 2235-00-9
Molecular Formula C8H13NO
Molecular Weight (g/mol) 139.198
MDL Number MFCD00080693
InChI Key JWYVGKFDLWWQJX-UHFFFAOYSA-N
Synonym n-vinylcaprolactam, 1-vinylazepan-2-one, n-vinyl-epsilon-caprolactam, 2h-azepin-2-one, 1-ethenylhexahydro, unii-kfc10cy9up, 1-vinylhexahydro-2h-azepin-2-one, 1-ethenylhexahydro-2h-azepin-2-one, kfc10cy9up, 1-vinylazaperhydroepin-2-one, poly-n-vinylcaprolactam
PubChem CID 75227
IUPAC Name 1-ethenylazepan-2-one
SMILES C=CN1CCCCCC1=O

Specifications

Melting Point 32°C to 36°C
Density 1.029
Boiling Point 128°C (21 mmHg)
Flash Point 101°C (214°F)
Quantity 10 g
Solubility Information Soluble in water (partly).
Formula Weight 139.2
Percent Purity 99%
Chemical Name or Material N-Vinyl-epsilon-caprolactam
Hazard Category H302-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280i-P301+P312-P304+P340-P305+P351+P338-P312-P330-P501c
EINECSNumber 218-787-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only