Learn More
n-Undecanoyl chloride, 98%
CAS: 17746-05-3 | C11H21ClO | 204.738 g/mol
$204.78 - $328.34
Chemical Identifiers
| CAS | 17746-05-3 |
|---|---|
| Molecular Formula | C11H21ClO |
| Molecular Weight (g/mol) | 204.738 |
| MDL Number | MFCD00000739 |
| InChI Key | JUKPJGZUFHCZQI-UHFFFAOYSA-N |
| Synonym | n-undecanoyl chloride, undecanoic acid chloride, undecanoylchloride, n-undecanoylchloride, undecanoyl chloride, acmc-1c80a, ksc175q6f |
| PubChem CID | 87287 |
| IUPAC Name | undecanoyl chloride |
| SMILES | CCCCCCCCCCC(=O)Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH56380ME
|
Thermo Scientific Chemicals
H56380ME |
500 mg |
N/A
|
|
|||||
|
AAH5638003
|
Thermo Scientific Chemicals
H5638003 |
1 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 17746-05-3 | |
| 204.738 | |
| JUKPJGZUFHCZQI-UHFFFAOYSA-N | |
| 87287 | |
| CCCCCCCCCCC(=O)Cl |
| C11H21ClO | |
| MFCD00000739 | |
| n-undecanoyl chloride, undecanoic acid chloride, undecanoylchloride, n-undecanoylchloride, undecanoyl chloride, acmc-1c80a, ksc175q6f | |
| undecanoyl chloride |
Specifications
| 17746-05-3 | |
| 113°C (235°F) | |
| 1.443 | |
| MFCD00000739 | |
| UN3265 | |
| Moisture sensitive | |
| JUKPJGZUFHCZQI-UHFFFAOYSA-N | |
| undecanoyl chloride | |
| 87287 | |
| 98% |
| 135°C to 136°C | |
| C11H21ClO | |
| CH3(CH2)9COCl | |
| 500 mg | |
| 1759226 | |
| n-undecanoyl chloride, undecanoic acid chloride, undecanoylchloride, n-undecanoylchloride, undecanoyl chloride, acmc-1c80a, ksc175q6f | |
| CCCCCCCCCCC(=O)Cl | |
| 204.738 | |
| 204.74 | |
| n-Undecanoyl chloride |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Hazard Class: 8; Packaging Group: II
EINECSNumber : 241-741-1
TSCA : Yes
Recommended Storage : Store at -20°C; Store under Nitrogen
RUO – Research Use Only