Learn More
N-(Trimethylsilyl)dimethylamine, 95%
CAS: 2083-91-2 | C5H15NSi | 117.27 g/mol
Supplier: Thermo Scientific Chemicals A1655018
Description
N,N-Dimethyltrimethylsilylamine was used in the synthesis of phosphoramidite. It was also employed as a reagent in the preparation of iminium salts and amides as well as for the silylation of polymers. A combination of N-(trimethylsilyl)dimethylamine and MeI was also effective to give p-methylbenzoic acid with a 85% yield based on 90% conversion from the corresponding Me ester. DETA has been treated with N-(trimethylsilyl)-dimethylamine to yield mono-, di- and tri- silylated derivatives
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN,N-Dimethyltrimethylsilylamine was used in the synthesis of phosphoramidite. It was also employed as a reagent in the preparation of iminium salts and amides as well as for the silylation of polymers. A combination of N-(trimethylsilyl)dimethylamine and MeI was also effective to give p-methylbenzoic acid with a 85% yield based on 90% conversion from the corresponding Me ester. DETA has been treated with N-(trimethylsilyl)-dimethylamine to yield mono-, di- and tri- silylated derivatives
Solubility
Hydrolyzes in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.
Specifications
| 86°C to 87°C | |
| 2083-91-2 | |
| C5H15NSi | |
| MFCD00008297 | |
| 1731861 | |
| n,n-dimethyltrimethylsilylamine, n,n-dimethylaminotrimethylsilane, silanamine, pentamethyl, dimethylamino trimethylsilane, pentamethylsilylamine, n-trimethylsilyl dimethylamine, tmsdma, dimethylaminotrimethylsilane, silanamine, n,n,1,1,1-pentamethyl, n,n-dimethyltrimethylsilamine | |
| KAHVZNKZQFSBFW-UHFFFAOYSA-N | |
| N-methyl-N-trimethylsilylmethanamine | |
| 74965 | |
| 95% | |
| 1.397 | |
| 0.732 |
| N-(Trimethylsilyl)dimethylamine | |
| 50 g | |
| (CH3)3SiN(CH3)2 | |
| UN2733 | |
| −19°C (−3°F) | |
| Hydrolyzes in water. | |
| CN(C)[Si](C)(C)C | |
| 117.27 | |
| 117.27 | |
| Amine-like | |
| Moisture sensitive |
Chemical Identifiers
| 2083-91-2 | |
| 117.27 | |
| KAHVZNKZQFSBFW-UHFFFAOYSA-N | |
| 74965 | |
| CN(C)[Si](C)(C)C |
| C5H15NSi | |
| MFCD00008297 | |
| n,n-dimethyltrimethylsilylamine, n,n-dimethylaminotrimethylsilane, silanamine, pentamethyl, dimethylamino trimethylsilane, pentamethylsilylamine, n-trimethylsilyl dimethylamine, tmsdma, dimethylaminotrimethylsilane, silanamine, n,n,1,1,1-pentamethyl, n,n-dimethyltrimethylsilamine | |
| N-methyl-N-trimethylsilylmethanamine |
Safety and Handling
GHS H Statement
H225-H314-H318
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H225-H314-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
EINECSNumber : 218-222-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only