accessibility menu, dialog, popup

UserName

H3CNCH3SiCH3CH3H3C

CAS RN 2083-91-2

IUPAC Name: dimethyl(trimethylsilyl)amine
Synonyms: dimethylamino-trimethyl silane
Molecular Weight (g/mol): 117.27
Molecular Formula: C5H15NSi
InChi Key: KAHVZNKZQFSBFW-UHFFFAOYSA-N
SMILES: CN(C)[Si](C)(C)C
Percent Purity 
  • (1)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (4)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)