missing translation for 'onlineSavingsMsg'
Learn More

n-Tributyl Phosphate, 96.5%, Spectrum™ Chemical

Catalog No. 18612055
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
Packaging:
Amber Glass Bottle
Steel Can
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18612055 20 L Steel Can
18612058 500 mL Amber Glass Bottle
18612057 4 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18612055 Supplier Spectrum Chemical Mfg Cor Supplier No. T110020LT
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 126-73-8

Specifications

CAS Max % 1
Packaging Steel Can
Quantity 20 L
Percent Purity 0.965
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.