missing translation for 'onlineSavingsMsg'
Learn More

n-Triacontane, 98+%

Catalog No. AAL0538606
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0538606 5 g
AAL05386MD 250 mg
AAL0538603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0538606 Supplier Thermo Scientific Chemicals Supplier No. L0538606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 638-68-6 | C30H62 | 422.826 g/mol

n-Triacontane is used to improve the threshold voltage and mobility for electrons of ambipolar organic light emitting field effect transistors with modified asymmetric electrodes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-Triacontane is used to improve the threshold voltage and mobility for electrons of ambipolar organic light emitting field effect transistors with modified asymmetric electrodes.

Solubility
Soluble in benzene, chloroform and toluene. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 638-68-6
Molecular Formula C30H62
Molecular Weight (g/mol) 422.826
MDL Number MFCD00009410
InChI Key JXTPJDDICSTXJX-UHFFFAOYSA-N
Synonym n-triacontane, triacontane, analytical standard, triacontane, acmc-1b4uf, n-triacontane 250mg, ch3-ch2 28-ch3
PubChem CID 12535
ChEBI CHEBI:31006
IUPAC Name triacontane
SMILES CCCCCCCCCCCCCCCCCCCCCCCCCCCCCC

Specifications

Melting Point 65°C to 68°C
Boiling Point 455°C
Flash Point 237°C (458°F)
Odor Odorless
Quantity 5 g
Beilstein 1777281
Solubility Information Soluble in benzene,chloroform and toluene. Insoluble in water.
Formula Weight 422.83
Percent Purity 99%
Chemical Name or Material n-Triacontane
EINECSNumber 211-349-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only