missing translation for 'onlineSavingsMsg'
Learn More

N-trans-Cinnamyl-N-methyl-(1-naphthylmethyl)amine Hydrochloride 98.0+%, TCI America™

Catalog No. C11625G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C11625G 5 g
C11621G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C11625G Supplier TCI America Supplier No. C11625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 65473-14-5
Molecular Formula C21H22ClN
Molecular Weight (g/mol) 323.864
MDL Number MFCD00059047
InChI Key OLUNPKFOFGZHRT-YGCVIUNWSA-N
Synonym naftifine hydrochloride, naftifine hcl, naftin, exoderil, naftifungin, n-trans-cinnamyl-n-methyl-1-naphthylmethyl amine hydrochloride, e-n-cinnamyl-n-methyl-1-naphthalenemethylamine hydrochloride, naftin tn, e-n-methyl-n-1-naphthylmethyl-3-phenyl-2-propen-1-amine-hydrochloride
PubChem CID 5281098
ChEBI CHEBI:7452
IUPAC Name (E)-N-methyl-N-(naphthalen-1-ylmethyl)-3-phenylprop-2-en-1-amine;hydrochloride
SMILES CN(CC=CC1=CC=CC=C1)CC2=CC=CC3=CC=CC=C32.Cl

Specifications

Melting Point 177°C
Color White-Yellow
Quantity 5 g
Formula Weight 323.86
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-trans-Cinnamyl-N-methyl-(1-naphthylmethyl)amine Hydrochloride
RTECSNumber QJ8650000
TSCA No