missing translation for 'onlineSavingsMsg'
Learn More

N-Toluenesulfonyl-L-proline, 94%

Catalog No. AAH6690214
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6690214 25 g
AAH6690206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6690214 Supplier Thermo Scientific Chemicals Supplier No. H6690214
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 51077-01-1 | C12H15NO4S | 269.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 51077-01-1
Molecular Formula C12H15NO4S
Molecular Weight (g/mol) 269.32
MDL Number MFCD00238392,MFCD00447113
InChI Key CGPHGPCHVUSFFA-UHFFFAOYNA-N
Synonym n-tosyl-l-proline, tos-pro-oh, 1-tosyl-l-proline, 2s-1-4-methylphenyl sulfonyl pyrrolidine-2-carboxylic acid, 2s-1-4-methylbenzenesulfonyl pyrrolidine-2-carboxylic acid, d,l-n-p-toluenesulfonyl proline, n-tosyl-proline, 1-4-methylphenyl sulfonyl proline #, proline, 1-4-methylphenyl sulfonyl, n-tosyl-s-proline
PubChem CID 691957
IUPAC Name 1-(4-methylbenzenesulfonyl)pyrrolidine-2-carboxylic acid
SMILES CC1=CC=C(C=C1)S(=O)(=O)N1CCCC1C(O)=O

Specifications

UN Number UN2811
Quantity 25 g
Formula Weight 269.32
Percent Purity 95%
Physical Form Low Melting Solid
Chemical Name or Material N-Toluenesulfonyl-L-proline
Hazard Category H301
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P301+P310-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only