accessibility menu, dialog, popup

UserName

CAS RN 51077-01-1

IUPAC Name: 1-(4-methylbenzenesulfonyl)pyrrolidine-2-carboxylic acid
Synonyms: tosyl-D-proline
Molecular Weight (g/mol): 269.32
Molecular Formula: C12H15NO4S
InChi Key: CGPHGPCHVUSFFA-UHFFFAOYNA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)N1CCCC1C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)