missing translation for 'onlineSavingsMsg'
Learn More

n-Tetradecane, EMD-Millipore

Catalog No. M1096080005
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1096080005 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1096080005 Supplier Technidata Supplier No. 1.09608.0005

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Hydrocarbons C9 – C19

Chemical Identifiers

CAS 629-59-4
Molecular Formula C14H30
Molecular Weight (g/mol) 198.394
InChI Key BGHCVCJVXZWKCC-UHFFFAOYSA-N
PubChem CID 12389
ChEBI CHEBI:41253
IUPAC Name tetradecane
SMILES CCCCCCCCCCCCCC

Specifications

Packaging Glass ampoule
Explosion Limit 0.5 to 6.5 %(V)
Ignition Point 202°C
Vapor Pressure 1.55 Pa (25°C)
Viscosity 3mm2/sec., kinematic (40°C)
Density Density: 0.76g/cm3 (20°)
Assay Percent Range ≥99.0%
Quantity 5 mL
Formula Weight 198.39
Chemical Name or Material n-Tetradecane
Recommended Storage Store without restrictions