missing translation for 'onlineSavingsMsg'
Learn More

n-Tetradecane, 99%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor T200720LTBL

 View more versions of this product

Catalog No. 18-612-052

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

629-59-4
Steel Can
0.99
1
20 L
Reagent

Chemical Identifiers

629-59-4
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.