missing translation for 'onlineSavingsMsg'
Learn More

n-Tetradecane, 99%, Spectrum™ Chemical

Catalog No. p-7166679
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Glass Bottle
Steel Can
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18612050 100 mL Amber Glass Bottle
18612054 500 mL Amber Glass Bottle
18612051 2.5 L Glass Bottle
18612052 20 L Steel Can
18612053 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18612050 Supplier Spectrum Chemical Mfg Cor Supplier No. T2007100ML
Only null left

Chemical Identifiers

CAS 629-59-4

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 100 mL
Percent Purity 0.99
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.