missing translation for 'onlineSavingsMsg'
Learn More

n-Tetracosane, 99%

Catalog No. AA4246206
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4246206 5 g
AA4246214 25 g
AA4246222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4246206 Supplier Thermo Scientific Chemicals Supplier No. 04246206

CAS: 646-31-1 | C24H50 | 338.664 g/mol

n-Tetracosane is a suitable solvent used in the synthesis of ZnS nanoparticle. It is also employed as the wax component to study the reduction in pour point of hydrocarbon solvents containing wax crystals on addition of polymer additives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 646-31-1
Molecular Formula C24H50
Molecular Weight (g/mol) 338.664
MDL Number MFCD00009352
InChI Key POOSGDOYLQNASK-UHFFFAOYSA-N
Synonym n-tetracosane, lignocerane, unii-yq5h1m1d7i, tricosane, methyl, yq5h1m1d7i, tetracosane, analytical standard, tetracosane, acmc-1b0gq, n-tetracosane 5g, ksc357o9b
PubChem CID 12592
ChEBI CHEBI:32936
IUPAC Name tetracosane
SMILES CCCCCCCCCCCCCCCCCCCCCCCC

Specifications

Melting Point 49°C to 52°C
Density 0.799
Boiling Point 391°C
Flash Point >110°C (230°F)
Quantity 5 g
Solubility Information Insoluble in water.
Formula Weight 338.66
Percent Purity 99%
Physical Form Crystalline
Chemical Name or Material n-Tetracosane
EINECSNumber 211-474-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only