missing translation for 'onlineSavingsMsg'
Learn More

N-tert-Butylisopropylamine, 98%

Catalog No. AAA1722106
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1722106 5 g
AAA1722114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1722106 Supplier Thermo Scientific Chemicals Supplier No. A1722106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7515-80-2 | C7H18N | 116.23 g/mol

N-tert-Butylisopropylamine, is used as a participates in the synthesis of hindered enamines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-tert-Butylisopropylamine, is used as a participates in the synthesis of hindered enamines.

Solubility
It has optimal solubility in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from oxidizing agents.

Chemical Identifiers

CAS 7515-80-2
Molecular Formula C7H18N
Molecular Weight (g/mol) 116.23
MDL Number MFCD00037059
InChI Key ZWXQPERWRDHCMZ-UHFFFAOYSA-O
Synonym n-tert-butylisopropylamine, 2-propanamine, 2-methyl-n-1-methylethyl, tert-butylisopropylamine, n-isopropyl-2-methylpropan-2-amine, ethanamine, 1,1-dimethyl, n-1-methylethyl, n-isopropyl-tert-butylamine, tert-butyl methylethyl amine, tert-butyl isopropyl amine
PubChem CID 82023
IUPAC Name 2-methyl-N-propan-2-ylpropan-2-amine
SMILES CC(C)[NH2+]C(C)(C)C

Specifications

Density 0.727
Boiling Point 98°C
Flash Point −3°C (26°F)
Refractive Index 1.398
Quantity 5 g
UN Number UN2733
Beilstein 1846752
Solubility Information It has optimal solubility in water.
Formula Weight 115.22
Percent Purity 98%
Chemical Name or Material N-tert-Butylisopropylamine
Show More Show Less
Hazard Category H225-H314
Hazard Statement GHS H Statement
H225-H314-H318
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
EINECSNumber 231-370-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only