accessibility menu, dialog, popup

UserName

CH3H3CNH2CH3CH3H3C

CAS RN 7515-80-2

IUPAC Name: tert-butyl(propan-2-yl)azanium
Synonyms: tert-butyl(isopropyl)azanium
Molecular Weight (g/mol): 116.23
Molecular Formula: C7H18N
InChi Key: ZWXQPERWRDHCMZ-UHFFFAOYSA-O
SMILES: CC(C)[NH2+]C(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results