missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-tert-Butylbenzylamine, TCI America™

$45.83 - $458.34

Chemical Identifiers

CAS 3378-72-1
Molecular Formula C11H17N
Molecular Weight (g/mol) 163.264
MDL Number MFCD00008798
InChI Key DLSOILHAKCBARI-UHFFFAOYSA-N
Synonym n-tert-butyl benzylamine, n-tert-butylbenzylamine, n-benzyl-tert-butylamine, tert-butylbenzylamine, n-t-butylbenzylamine, benzenemethanamine, n-1,1-dimethylethyl, benzyl tert-butyl amine, tert-butyl benzylamine, t-butylbenzylamine, n-1,1-dimethylethyl benzylamine
PubChem CID 76908
IUPAC Name N-benzyl-2-methylpropan-2-amine
SMILES CC(C)(C)NCC1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B117625ML View Documents TCI America
B117625ML
25mL Each for $45.83
 
B1176500ML View Documents TCI America
B1176500ML
500mL Each for $458.34
 
Specifications

Chemical Identifiers

3378-72-1
163.264
DLSOILHAKCBARI-UHFFFAOYSA-N
76908
CC(C)(C)NCC1=CC=CC=C1
C11H17N
MFCD00008798
n-tert-butyl benzylamine, n-tert-butylbenzylamine, n-benzyl-tert-butylamine, tert-butylbenzylamine, n-t-butylbenzylamine, benzenemethanamine, n-1,1-dimethylethyl, benzyl tert-butyl amine, tert-butyl benzylamine, t-butylbenzylamine, n-1,1-dimethylethyl benzylamine
N-benzyl-2-methylpropan-2-amine

Specifications

252°C
25mL
C11H17N
2735
DLSOILHAKCBARI-UHFFFAOYSA-N
N-benzyl-2-methylpropan-2-amine
76908
Clear Liquid at 20°C
N-tert-Butylbenzylamine
Yellow
3378-72-1
MFCD00008798
n-tert-butyl benzylamine, n-tert-butylbenzylamine, n-benzyl-tert-butylamine, tert-butylbenzylamine, n-t-butylbenzylamine, benzenemethanamine, n-1,1-dimethylethyl, benzyl tert-butyl amine, tert-butyl benzylamine, t-butylbenzylamine, n-1,1-dimethylethyl benzylamine
CC(C)(C)NCC1=CC=CC=C1
163.264
163.26
≥96.0% (GC)
Safety and Handling

Safety and Handling

TSCA : Yes