missing translation for 'onlineSavingsMsg'
Learn More

N-tert-Butylbenzylamine 96.0+%, TCI America™

Catalog No. B117625ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B117625ML 25 mL
B1176500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B117625ML Supplier TCI America Supplier No. B117625ML

Chemical Identifiers

CAS 3378-72-1
Molecular Formula C11H17N
Molecular Weight (g/mol) 163.264
MDL Number MFCD00008798
InChI Key DLSOILHAKCBARI-UHFFFAOYSA-N
Synonym n-tert-butyl benzylamine, n-tert-butylbenzylamine, n-benzyl-tert-butylamine, tert-butylbenzylamine, n-t-butylbenzylamine, benzenemethanamine, n-1,1-dimethylethyl, benzyl tert-butyl amine, tert-butyl benzylamine, t-butylbenzylamine, n-1,1-dimethylethyl benzylamine
PubChem CID 76908
IUPAC Name N-benzyl-2-methylpropan-2-amine
SMILES CC(C)(C)NCC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 252°C
Quantity 25 mL
UN Number 2735
Formula Weight 163.26
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material N-tert-Butylbenzylamine