missing translation for 'onlineSavingsMsg'
Learn More

N-tert-Butyl-alpha-(4-pyridyl-1-oxide)nitrone 98.0+%, TCI America™

Catalog No. p-7119232
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B17351G 1 g
B17355G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B17351G Supplier TCI America Supplier No. B17351G

Chemical Identifiers

CAS 66893-81-0
Molecular Formula C10H14N2O2
Molecular Weight (g/mol) 194.23
MDL Number MFCD00041022
InChI Key RNRMWTCECDHNQU-UHFFFAOYSA-N
Synonym pobn, n-tert-butyl-alpha-4-pyridyl-1-oxide nitrone, unii-2140sm8q5o, alpha-4-pyridyl-1-oxide-n-tert-butylnitrone, pyridyloxide tert-butylnitrone, 4-tert-butyl oxo ammonio methylidene pyridin-1 4h-olate, alpha-4-pyridyl-n-oxide-n-t-butylnitrone, alpha-4-pyridyl n-oxide-n-tert-butylnitrone, n-tert-butyl-alpha-4-pyridyl nitrone n'-oxide, alpha-4-pyridyl-1-oxide-n-t-butylnitrone
PubChem CID 92375
IUPAC Name 4-[(tert-butyl-oxo-λ⁵-azanylidene)methyl]pyridin-1-ium-1-olate
SMILES CC(C)(C)[N+]([O-])=CC1=CC=[N+]([O-])C=C1

Specifications

Melting Point 185°C
Color White-Yellow
Quantity 1 g
Formula Weight 194.23
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-tert-Butyl-alpha-(4-pyridyl-1-oxide)nitrone
TSCA No
Recommended Storage Refrigerator