accessibility menu, dialog, popup

UserName

CH3H3CCH3NONO(E/Z)

CAS RN 66893-81-0

IUPAC Name: 4-[(tert-butyl-oxo-λ⁵-azanylidene)methyl]pyridin-1-ium-1-olate
Synonyms: pyridyloxide T-butylnitrone
Molecular Weight (g/mol): 194.23
Molecular Formula: C10H14N2O2
InChi Key: RNRMWTCECDHNQU-UHFFFAOYSA-N
SMILES: CC(C)(C)[N+]([O-])=CC1=CC=[N+]([O-])C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results