missing translation for 'onlineSavingsMsg'
Learn More

N-tert-Butyl-3,5-dimethylaniline 98.0+%, TCI America™

Catalog No. p-7119834
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B29031G 1 g
B29035G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B29031G Supplier TCI America Supplier No. B29031G
Only null left

Chemical Identifiers

CAS 110993-40-3
Molecular Formula C12H19N
Molecular Weight (g/mol) 177.291
InChI Key HMWHVIXDKZHDEO-UHFFFAOYSA-N
Synonym n-tert-butyl-3,5-dimethylaniline, n-tert-butyl-3,5-xylidine, 3,5-dimethyl-n-tert-butylaniline, n-1,1-dimethylethyl-3,5-dimethyl benzenamine, benzenamine,n-1,1-dimethylethyl-3,5-dimethyl
PubChem CID 5242234
IUPAC Name N-tert-butyl-3,5-dimethylaniline
SMILES CC1=CC(=CC(=C1)NC(C)(C)C)C

Specifications

Boiling Point 70°C
Quantity 1 g
Physical Form Clear Liquid at 20°C
Formula Weight 177.29
Percent Purity ≥98.0% (GC)
Chemical Name or Material N-tert-Butyl-3,5-dimethylaniline
TSCA Yes
Recommended Storage Refrigerator