missing translation for 'onlineSavingsMsg'
Learn More

N-tert-Butyl-2-benzothiazolesulfenamide, 97%

Catalog No. AAL1335630
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1335630 250 g
AAL1335618 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1335630 Supplier Thermo Scientific Chemicals Supplier No. L1335630
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 95-31-8 | C11H14N2S2 | 238.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 95-31-8
Molecular Formula C11H14N2S2
Molecular Weight (g/mol) 238.37
MDL Number MFCD00022873
InChI Key IUJLOAKJZQBENM-UHFFFAOYSA-N
Synonym n-tert-butyl-2-benzothiazolesulfenamide, santocure ns, nocceler ns, vulkacit nz, accel bns, pennac tbbs, vanax ns, s-benzo d thiazol-2-yl-n-tert-butyl thiohydroxylamine, 2-benzothiazolesulfenamide, n-1,1-dimethylethyl, 2-benzothiazolesulfenamide, n-tert-butyl
PubChem CID 7230
IUPAC Name N-(1,3-benzothiazol-2-ylsulfanyl)-2-methylpropan-2-amine
SMILES CC(C)(C)NSC1=NC2=CC=CC=C2S1

Specifications

Melting Point 105°C to 110°C
Density 1.29
Flash Point 165°C (329°F)
Quantity 250 g
Beilstein 158370
Formula Weight 238.38
Percent Purity 97%
Chemical Name or Material N-tert-Butyl-2-benzothiazolesulfenamide
Hazard Category H317
Hazard Statement GHS H Statement
H317
May cause an allergic skin reaction.
Precautionary Statement P261-P272-P280g-P302+P352-P333+P313-P363-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 202-409-1
RTECSNumber DL6200000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only