missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)-O-benzyl-L-tyrosine 98.0+%, TCI America™

Catalog No. p-7119170
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B16315G 5 g
B163125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B16315G Supplier TCI America Supplier No. B16315G
Only null left

Chemical Identifiers

CAS 2130-96-3
Molecular Formula C21H25NO5
Molecular Weight (g/mol) 371.43
MDL Number MFCD00065597
InChI Key ZAVSPTOJKOFMTA-LGWFVXIRNA-N
Synonym boc-tyr bzl-oh, boc-o-benzyl-l-tyrosine, n-boc-o-benzyl-l-tyrosine, n-tert-butoxycarbonyl-o-benzyl-l-tyrosine, s-3-4-benzyloxy phenyl-2-tert-butoxycarbonyl amino propanoic acid, n-tert-butoxycarbonyl-o-benzyl-l-tyrisine, o-benzyl-n-tert-butoxycarbonyl-l-tyrosine, 2s-3-4-benzyloxy phenyl-2-tert-butoxycarbonyl amino propanoic acid, o-benzyl-n-tert-butoxy carbonyl-l-tyrosine, tyrosine, n-1,1-dimethylethoxy carbonyl-o-phenylmethyl
PubChem CID 98763
IUPAC Name (2S)-3-[4-(benzyloxy)phenyl]-2-{[(tert-butoxy)carbonyl]amino}propanoic acid
SMILES CC(C)(C)OC(=O)N[C@@H](CC1=CC=C(OCC2=CC=CC=C2)C=C1)C(O)=O

Specifications

Melting Point 113°C
Color White-Yellow
Quantity 5 g
Formula Weight 371.43
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-O-benzyl-L-tyrosine