accessibility menu, dialog, popup

UserName

HN(S)OOOOHOCH3CH3CH3

CAS RN 2130-96-3

IUPAC Name: (2S)-3-[4-(benzyloxy)phenyl]-2-{[(tert-butoxy)carbonyl]amino}propanoic acid
Synonyms: (2S)-3-[4-(benzyloxy)phenyl]-2-[(tert-butoxycarbonyl)amino]propanoic acid
Molecular Weight (g/mol): 371.43
Molecular Formula: C21H25NO5
InChi Key: ZAVSPTOJKOFMTA-LGWFVXIRNA-N
SMILES: CC(C)(C)OC(=O)N[C@@H](CC1=CC=C(OCC2=CC=CC=C2)C=C1)C(O)=O
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)