missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)-O-benzyl-L-threonine 98.0+%, TCI America™

Catalog No. b16305g
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B16305G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B16305G Supplier TCI America Supplier No. B16305G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 15260-10-3
Molecular Formula C16H23NO5
Molecular Weight (g/mol) 309.36
MDL Number MFCD00066062
InChI Key CTXPLTPDOISPTE-YPMHNXCESA-N
Synonym boc-thr bzl-oh, boc-o-benzyl-l-threonine, n-tert-butoxycarbonyl-o-benzyl-l-threonine, n-boc-o-benzyl-l-threonine, boc-thr bzl, o-benzyl-n-tert-butoxycarbonyl-l-threonine, n-1,1-dimethylethoxy carbonyl-o-phenylmethyl-l-threonine, l-threonine, n-1,1-dimethylethoxy carbonyl-o-phenylmethyl, 2s,3r-2-2-methylpropan-2-yl oxycarbonylamino-3-phenylmethoxybutanoic acid
PubChem CID 1549483
IUPAC Name (2S,3R)-3-(benzyloxy)-2-{[(tert-butoxy)carbonyl]amino}butanoic acid
SMILES C[C@@H](OCC1=CC=CC=C1)[C@H](NC(=O)OC(C)(C)C)C(O)=O

Spécifications

Melting Point 116°C
Color White
Quantity 5 g
Formula Weight 309.36
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-O-benzyl-L-threonine