missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)-O-benzyl-L-serine 98.0+%, TCI America™

Catalog No. B16295G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B16295G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B16295G Supplier TCI America Supplier No. B16295G

Chemical Identifiers

CAS 23680-31-1
Molecular Formula C15H21NO5
Molecular Weight (g/mol) 295.335
MDL Number MFCD00066063
InChI Key DMBKPDOAQVGTST-LBPRGKRZSA-N
Synonym boc-ser bzl-oh, n-boc-o-benzyl-l-serine, boc-o-benzyl-l-serine, boc-ser bzl, s-3-benzyloxy-2-tert-butoxycarbonyl amino propanoic acid, n-1,1-dimethylethoxy carbonyl-o-phenylmethyl-l-serine, n-t-butoxycarbonyl-o-benzyl-l-serine, o-benzyl-n-tert-butyloxycarbonylserine, n-tert-butoxycarbonyl-o-benzyl-l-serine, o-benzyl-n-tert-butoxycarbonyl-l-serine
PubChem CID 90234
IUPAC Name (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-phenylmethoxypropanoic acid
SMILES CC(C)(C)OC(=O)NC(COCC1=CC=CC=C1)C(=O)O

Specifications

Melting Point 59°C
Color White
Quantity 5 g
Formula Weight 295.34
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-O-benzyl-L-serine