missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(tert-Butoxycarbonyl)-N-methyl-1,2-ethylenediamine 95.0+%, TCI America™
$82.02 - $246.05
Chemical Identifiers
| CAS | 121492-06-6 |
|---|---|
| Molecular Formula | C8H18N2O2 |
| Molecular Weight (g/mol) | 174.244 |
| MDL Number | MFCD01317789 |
| InChI Key | QYJVBVKFXDHFPQ-UHFFFAOYSA-N |
| Synonym | n-boc-n-methylethylenediamine, n-2-aminoethyl-n-methyl carbamic acid tert-butyl ester, n-boc-2-aminoethyl-n-methylamine, tert-butyl n-2-aminoethyl-n-methylcarbamate, n-boc-n-methyl-ethylenediamine, tert-butyl 2-aminoethyl methyl carbamate, 2-n-boc-n-methylamino ethylamine, n-2-aminoethyl-n-methylcarbamicacidtert-butylester, n-boc-n-methyl ethylenediamine |
| PubChem CID | 2756424 |
| IUPAC Name | tert-butyl N-(2-aminoethyl)-N-methylcarbamate |
| SMILES | CC(C)(C)OC(=O)N(C)CCN |
Chemical Identifiers
| 121492-06-6 | |
| 174.244 | |
| QYJVBVKFXDHFPQ-UHFFFAOYSA-N | |
| 2756424 | |
| CC(C)(C)OC(=O)N(C)CCN |
| C8H18N2O2 | |
| MFCD01317789 | |
| n-boc-n-methylethylenediamine, n-2-aminoethyl-n-methyl carbamic acid tert-butyl ester, n-boc-2-aminoethyl-n-methylamine, tert-butyl n-2-aminoethyl-n-methylcarbamate, n-boc-n-methyl-ethylenediamine, tert-butyl 2-aminoethyl methyl carbamate, 2-n-boc-n-methylamino ethylamine, n-2-aminoethyl-n-methylcarbamicacidtert-butylester, n-boc-n-methyl ethylenediamine | |
| tert-butyl N-(2-aminoethyl)-N-methylcarbamate |
Specifications
| 121492-06-6 | |
| Colorless | |
| MFCD01317789 | |
| n-boc-n-methylethylenediamine, n-2-aminoethyl-n-methyl carbamic acid tert-butyl ester, n-boc-2-aminoethyl-n-methylamine, tert-butyl n-2-aminoethyl-n-methylcarbamate, n-boc-n-methyl-ethylenediamine, tert-butyl 2-aminoethyl methyl carbamate, 2-n-boc-n-methylamino ethylamine, n-2-aminoethyl-n-methylcarbamicacidtert-butylester, n-boc-n-methyl ethylenediamine | |
| CC(C)(C)OC(=O)N(C)CCN | |
| 1 mL | |
| 2756424 | |
| ≥95.0% (GC,T) | |
| N-(tert-Butoxycarbonyl)-N-methyl-1,2-ethylenediamine |
| 79°C | |
| C8H18N2O2 | |
| 2735 | |
| QYJVBVKFXDHFPQ-UHFFFAOYSA-N | |
| tert-butyl N-(2-aminoethyl)-N-methylcarbamate | |
| 174.244 | |
| 174.24 | |
| Liquid |
Safety and Handling
TSCA : No