accessibility menu, dialog, popup

UserName

CAS RN 121492-06-6

CH3CH3H3COONCH3H2N

CAS RN 121492-06-6

IUPAC Name: tert-butyl N-(2-aminoethyl)-N-methylcarbamate
Synonyms: tert-butyl N-(2-aminoethyl)-N-methylcarbamate
Molecular Weight (g/mol): 174.24
Molecular Formula: C8H18N2O2
InChi Key: QYJVBVKFXDHFPQ-UHFFFAOYSA-N
SMILES: CN(CCN)C(=O)OC(C)(C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)