missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)-L-threonine 98.0+%, TCI America™

Catalog No. B163810G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B163810G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B163810G Supplier TCI America Supplier No. B163810G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2592-18-9
Molecular Formula C9H17NO5
Molecular Weight (g/mol) 219.237
MDL Number MFCD00065946
InChI Key LLHOYOCAAURYRL-RITPCOANSA-N
Synonym boc-thr-oh, boc-l-threonine, n-boc-l-threonine, boc-threonine, n-tert-butoxycarbonyl-l-threonine, 2s,3r-2-tert-butoxycarbonyl amino-3-hydroxybutanoic acid, threonine, n-1,1-dimethylethoxy carbonyl, 2s,3r-2-tert-butoxycarbonylamino-3-hydroxybutanoic acid, 2s,3r-2-tert-butoxy carbonyl amino-3-hydroxybutanoic acid
PubChem CID 2724766
IUPAC Name (2S,3R)-3-hydroxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
SMILES CC(C(C(=O)O)NC(=O)OC(C)(C)C)O

Specifications

Melting Point 84°C
Color White
Quantity 10 g
Formula Weight 219.24
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-L-threonine