accessibility menu, dialog, popup

UserName

H3C(R)OH(S)NHOOCH3CH3H3COO

CAS RN 2592-18-9

IUPAC Name: (2S,3R)-2-{[(tert-butoxy)carbonyl]amino}-3-hydroxybutanoate
Synonyms: (2S,3R)-2-[(tert-butoxycarbonyl)amino]-3-hydroxybutanoate
Molecular Weight (g/mol): 218.23
Molecular Formula: C9H16NO5
InChi Key: LLHOYOCAAURYRL-RITPCOANSA-M
SMILES: C[C@@H](O)[C@H](NC(=O)OC(C)(C)C)C([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results