missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)-L-serine 97.0+%, TCI America™

Supplier:  TCI America B163725G

Quantity 25 g

 View more versions of this product

Catalog No. B163725G

Only null left

Description

Description

Specifications

Chemical Identifiers

3262-72-4
205.21
FHOAKXBXYSJBGX-MLHKIVSYNA-N
98766
CC(C)(C)OC(=O)N[C@@H](CO)C(O)=O
C8H15NO5
MFCD00037243
boc-ser-oh, n-boc-l-serine, boc-l-serine, n-tert-butoxycarbonyl-l-serine, n-boc-serine, s-n-boc-serine, n-tert-butoxycarbonyl serine, s-2-tert-butoxycarbonyl amino-3-hydroxypropanoic acid, n-t-butoxycarbonyl-l-serine, boc-l-ser
(2S)-2-{[(tert-butoxy)carbonyl]amino}-3-hydroxypropanoic acid

Specifications

N-(tert-Butoxycarbonyl)-L-serine
White
MFCD00037243
FHOAKXBXYSJBGX-MLHKIVSYNA-N
(2S)-2-{[(tert-butoxy)carbonyl]amino}-3-hydroxypropanoic acid
205.21
205.21
Crystalline Powder
3262-72-4
C8H15NO5
boc-ser-oh, n-boc-l-serine, boc-l-serine, n-tert-butoxycarbonyl-l-serine, n-boc-serine, s-n-boc-serine, n-tert-butoxycarbonyl serine, s-2-tert-butoxycarbonyl amino-3-hydroxypropanoic acid, n-t-butoxycarbonyl-l-serine, boc-l-ser
CC(C)(C)OC(=O)N[C@@H](CO)C(O)=O
25 g
98766
≥97.0% (T)
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions