accessibility menu, dialog, popup

UserName

(S)NHOOOOHHOH3CCH3H3C

CAS RN 3262-72-4

IUPAC Name: (2S)-2-{[(tert-butoxy)carbonyl]amino}-3-hydroxypropanoic acid
Synonyms: (2S)-2-[(tert-butoxycarbonyl)amino]-3-hydroxypropanoic acid|BOC-L-Serine hydrate
Molecular Weight (g/mol): 205.21
Molecular Formula: C8H15NO5
InChi Key: FHOAKXBXYSJBGX-MLHKIVSYNA-N
SMILES: CC(C)(C)OC(=O)N[C@@H](CO)C(O)=O
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (1)
  • (3)
  • (3)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (5)
  • (2)
  • (5)
  • (1)
  • (3)
  • (3)
  • (3)