missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(tert-Butoxycarbonyl)-L-proline 99.0+%, TCI America™
$35.41 - $102.52
Chemical Identifiers
| CAS | 15761-39-4 |
|---|---|
| Molecular Formula | C10H17NO4 |
| Molecular Weight (g/mol) | 215.249 |
| MDL Number | MFCD00037324 |
| InChI Key | ZQEBQGAAWMOMAI-ZETCQYMHSA-N |
| Synonym | boc-l-proline, boc-pro-oh, n-boc-l-proline, n-tert-butoxycarbonyl-l-proline, boc-l-pro-oh, s-1-tert-butoxycarbonyl pyrrolidine-2-carboxylic acid, n-tert-butyloxycarbonyl-l-proline, tert-butoxycarbonyl-l-proline, 1-tert-butoxycarbonyl-l-proline, boc-l-pro |
| PubChem CID | 85083 |
| IUPAC Name | (2S)-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-2-carboxylic acid |
| SMILES | CC(C)(C)OC(=O)N1CCCC1C(=O)O |
Chemical Identifiers
| 15761-39-4 | |
| 215.249 | |
| ZQEBQGAAWMOMAI-ZETCQYMHSA-N | |
| 85083 | |
| CC(C)(C)OC(=O)N1CCCC1C(=O)O |
| C10H17NO4 | |
| MFCD00037324 | |
| boc-l-proline, boc-pro-oh, n-boc-l-proline, n-tert-butoxycarbonyl-l-proline, boc-l-pro-oh, s-1-tert-butoxycarbonyl pyrrolidine-2-carboxylic acid, n-tert-butyloxycarbonyl-l-proline, tert-butoxycarbonyl-l-proline, 1-tert-butoxycarbonyl-l-proline, boc-l-pro | |
| (2S)-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-2-carboxylic acid |
Specifications
| 15761-39-4 | |
| White | |
| MFCD00037324 | |
| boc-l-proline, boc-pro-oh, n-boc-l-proline, n-tert-butoxycarbonyl-l-proline, boc-l-pro-oh, s-1-tert-butoxycarbonyl pyrrolidine-2-carboxylic acid, n-tert-butyloxycarbonyl-l-proline, tert-butoxycarbonyl-l-proline, 1-tert-butoxycarbonyl-l-proline, boc-l-pro | |
| CC(C)(C)OC(=O)N1CCCC1C(=O)O | |
| 215.249 | |
| 215.25 | |
| Crystalline Powder |
| 136°C | |
| C10H17NO4 | |
| 5 g | |
| ZQEBQGAAWMOMAI-ZETCQYMHSA-N | |
| (2S)-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-2-carboxylic acid | |
| 85083 | |
| ≥99.0% (T) | |
| N-(tert-Butoxycarbonyl)-L-proline |
Safety and Handling
TSCA : Yes