missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(tert-Butoxycarbonyl)-L-proline 99.0+%, TCI America™
Supplier: TCI America B118825G
Specifications
| N-(tert-Butoxycarbonyl)-L-proline | |
| 136°C | |
| C10H17NO4 | |
| 25 g | |
| ZQEBQGAAWMOMAI-ZETCQYMHSA-N | |
| (2S)-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-2-carboxylic acid | |
| 85083 | |
| ≥99.0% (T) |
| 15761-39-4 | |
| White | |
| MFCD00037324 | |
| boc-l-proline, boc-pro-oh, n-boc-l-proline, n-tert-butoxycarbonyl-l-proline, boc-l-pro-oh, s-1-tert-butoxycarbonyl pyrrolidine-2-carboxylic acid, n-tert-butyloxycarbonyl-l-proline, tert-butoxycarbonyl-l-proline, 1-tert-butoxycarbonyl-l-proline, boc-l-pro | |
| CC(C)(C)OC(=O)N1CCCC1C(=O)O | |
| 215.249 | |
| 215.25 | |
| Crystalline Powder |
Chemical Identifiers
| 15761-39-4 | |
| 215.249 | |
| ZQEBQGAAWMOMAI-ZETCQYMHSA-N | |
| 85083 | |
| CC(C)(C)OC(=O)N1CCCC1C(=O)O |
| C10H17NO4 | |
| MFCD00037324 | |
| boc-l-proline, boc-pro-oh, n-boc-l-proline, n-tert-butoxycarbonyl-l-proline, boc-l-pro-oh, s-1-tert-butoxycarbonyl pyrrolidine-2-carboxylic acid, n-tert-butyloxycarbonyl-l-proline, tert-butoxycarbonyl-l-proline, 1-tert-butoxycarbonyl-l-proline, boc-l-pro | |
| (2S)-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-2-carboxylic acid |
Safety and Handling
TSCA : Yes